missing translation for 'onlineSavingsMsg'
Learn More

O-(Tetrahydropyran-2-yl)hydroxylamine, 96%

Product Code. 11955661 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11955661

Brand: Thermo Scientific Acros 437850010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6723-30-4 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6723-30-4
CAS Min % 95.0
CAS Max % 100.0
Melting Point 32.7°C to 37.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C5H11NO2
MDL Number MFCD01321374
Quantity 1 g
Synonym o-tetrahydro-2h-pyran-2-yl hydroxylamine, o-tetrahydropyran-2-yl-hydroxylamine, o-oxan-2-yl hydroxylamine, o-tetrahydropyran-2-yl hydroxylamine, 2-aminooxy tetrahydro-2h-pyran, o-tetrahydro-pyran-2-yl-hydroxylamine, o-tetrahydropyran-2-ylhydroxylamine, o-tertrahydroxy-pyran-2-yl-hydroxylamine, hydroxylamine, o-tetrahydro-2h-pyran-2-yl, h2nothp
InChI Key NLXXVSKHVGDQAT-UHFFFAOYNA-N
SMILES NOC1CCCCO1
IUPAC Name O-(oxan-2-yl)hydroxylamine
Molecular Weight (g/mol) 117.15
PubChem CID 5142091
Formula Weight 117.15
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material O-(Tetrahydropyran-2-yl)hydroxylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.