Learn More
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
£36.68 - £1329.62
Chemical Identifiers
CAS | 612-82-8 |
---|---|
Molecular Formula | C14H16N2·2HCl |
Molecular Weight (g/mol) | 285.2 |
InChI Key | LUKPNZHXJRJBAN-UHFFFAOYSA-N |
Synonym | 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl |
PubChem CID | 108938 |
IUPAC Name | 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride |
SMILES | CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10581524
|
Thermo Scientific Acros
421130250 |
25 g | Glass Bottle |
£36.68
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10299600
|
Thermo Scientific Acros
421131000 |
100 g | Glass Bottle |
£94.67
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10015660
|
Thermo Scientific Acros
421135000 |
500 g | Glass Bottle |
£315.57
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935271
|
Thermo Scientific Acros
421130025 |
2.5 kg | Plastic bottle |
£1329.62
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
612-82-8 | |
285.2 | |
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride |
C14H16N2·2HCl | |
LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
108938 | |
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl |
Specifications
612-82-8 | |
Beige to Beige-Gray | |
98.5% min. (on dry substance) (Argentometry) | |
15,9675 | |
LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride | |
108938 | |
99% | |
o-Tolidine dihydrochloride |
130.0°C | |
Authentic | |
C14H16N2·2HCl | |
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl | |
285.2 | |
285.2 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-322-5
- o-Tolidine dihydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
Hazard Statement
- H302-Harmful if swallowed.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 3,3-Dimethylbenzidine dihydrochloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.