missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenethiol, 97%, Thermo Scientific Chemicals

Product Code. 10310644
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-06-4
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004838
InChI Key LXUNZSDDXMPKLP-UHFFFAOYSA-N
Synonym o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
PubChem CID 8712
IUPAC Name 2-methylbenzenethiol
SMILES CC1=CC=CC=C1S

Specifications

Melting Point 14.0°C
Color Colorless to Yellow
Density 1.0540g/mL
Boiling Point 194.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SH
Refractive Index 1.5790 to 1.5810
Quantity 5 g
Beilstein 06,37
Merck Index 15,9478
Specific Gravity 1.054
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 124.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material o-Toluenethiol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Toluenethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'