missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11409887
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-69-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004802
InChI Key JYKYYPPZLPVIBY-UHFFFAOYSA-N
Synonym 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate
PubChem CID 69195
IUPAC Name 1-isothiocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=S

Specifications

Density 1.115
Boiling Point 239°C
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.6361
Quantity 25 g
UN Number UN2922
Beilstein 878487
Sensitivity Moisture sensitive
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material o-Tolyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'