Learn More
o-Tolyl isothiocyanate, 98%, Thermo Scientific Chemicals
£32.20 - £293.00
Chemical Identifiers
CAS | 614-69-7 |
---|---|
Molecular Formula | C8H7NS |
Molecular Weight (g/mol) | 149.211 |
MDL Number | MFCD00004802 |
InChI Key | JYKYYPPZLPVIBY-UHFFFAOYSA-N |
Synonym | 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate |
PubChem CID | 69195 |
IUPAC Name | 1-isothiocyanato-2-methylbenzene |
SMILES | CC1=CC=CC=C1N=C=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499877
|
Thermo Scientific Alfa Aesar
B22845.06 |
5 g |
£32.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409887
|
Thermo Scientific Alfa Aesar
B22845.14 |
25 g |
£103.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419887
|
Thermo Scientific Alfa Aesar
B22845.22 |
100 g |
£293.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
614-69-7 | |
149.211 | |
JYKYYPPZLPVIBY-UHFFFAOYSA-N | |
69195 | |
CC1=CC=CC=C1N=C=S |
C8H7NS | |
MFCD00004802 | |
2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate | |
1-isothiocyanato-2-methylbenzene |
Specifications
614-69-7 | |
239°C | |
Pungent | |
1.6361 | |
UN2922 | |
Moisture sensitive | |
JYKYYPPZLPVIBY-UHFFFAOYSA-N | |
1-isothiocyanato-2-methylbenzene | |
69195 | |
98% |
1.115 | |
107°C (224°F) | |
C8H7NS | |
MFCD00004802 | |
878487 | |
2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate | |
CC1=CC=CC=C1N=C=S | |
149.211 | |
149.22 | |
o-Tolyl isothiocyanate |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P342+P311-P363-P501c
H302+H312+H332-H317-H334
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 210-390-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only