missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11452428
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-26-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD00012925
InChI Key KJGFNDCSTWGUDT-UHFFFAOYSA-N
Synonym o-tolylhydrazine hydrochloride, 2-methylphenyl hydrazine hydrochloride, 2-methylphenylhydrazine hydrochloride, o-tolylhydrazine hcl, 1-2-methylphenyl hydrazine hydrochloride, 2-tolylhydrazine hydrochloride, o-tolylhydrazinium 1+ chloride, o-methylphenylhydrazine hcl, 2-methylphenylhydrazine hcl, hydrazine, 2-methylphenyl-, monohydrochloride
PubChem CID 71554
IUPAC Name (2-methylphenyl)hydrazine;hydrochloride
SMILES CC1=CC=CC=C1NN.Cl

Specifications

Melting Point ∼190°C (decomposition)
Quantity 100 g
UN Number UN2811
Beilstein 3655929
Sensitivity Hygroscopic
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material o-Tolylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'