missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Xylene, Reagent Grade, ≥98.0%, Honeywell
Reagent Grade,≥98.0%
£31.76 - £634.17
Chemical Identifiers
CAS | 95-47-6 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008519 |
InChI Key | CTQNGGLPUBDAKN-UHFFFAOYSA-N |
Synonym | o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes |
PubChem CID | 7237 |
ChEBI | CHEBI:28063 |
IUPAC Name | 1,2-xylene |
SMILES | CC1=CC=CC=C1C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15658200
|
Honeywell Research Chemicals
X1040-1L |
1 L | Glass Bottle | N/A | |||||
15648200
|
Honeywell Research Chemicals
X1040-18L |
18 L | Stainless Steel Can |
£634.17
18 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
95-47-6 | |
106.17 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
7237 | |
1,2-xylene |
C8H10 | |
MFCD00008519 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CHEBI:28063 | |
CC1=CC=CC=C1C |
Specifications
-26°C to -23°C (literature) | |
100.0000% | |
143°C to 145°C | |
30°C | |
C6H5CF3 | |
16 mmHg (37.7°C), 0.1 atm (21.1°C), 7 mmHg (20°C) | |
1815558 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
7237 | |
106.17 | |
Reagent |
95-47-6 | |
0.879 g/mL at 20 °C(lit.) | |
0.07 | |
C8H10 | |
MFCD00008519 | |
UN1307 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CC1=CC=CC=C1C | |
106.17 | |
CHEBI:28063 | |
>98% | |
o-Xylene, Reagent Grade, ≥98.0% |
Safety and Handling
P210-P261-P273-P301 + P310-P302 + P352 + P312-P331
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Xylenes
missing translation for 'einecsNumber' : 202-422-2
missing translation for 'rtecsNumber' : 202-422-2
Recommended Storage : Room Temp
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : o-xylene