Learn More
o-Xylylene dibromide, 97%, Thermo Scientific Chemicals
£20.20 - £113.00
Chemical Identifiers
CAS | 91-13-4 |
---|---|
Molecular Formula | C8H8Br2 |
Molecular Weight (g/mol) | 263.96 |
MDL Number | MFCD00000175 |
InChI Key | KGKAYWMGPDWLQZ-UHFFFAOYSA-N |
Synonym | 1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776 |
PubChem CID | 66665 |
IUPAC Name | 1,2-bis(bromomethyl)benzene |
SMILES | C1=CC=C(C(=C1)CBr)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423467
|
Thermo Scientific Alfa Aesar
A17241.06 |
5 g |
£20.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433467
|
Thermo Scientific Alfa Aesar
A17241.14 |
25 g |
£39.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443467
|
Thermo Scientific Alfa Aesar
A17241.22 |
100 g |
£113.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.
Solubility
Solubility in toluene is almost transparent and ethanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
91-13-4 | |
263.96 | |
KGKAYWMGPDWLQZ-UHFFFAOYSA-N | |
66665 | |
C1=CC=C(C(=C1)CBr)CBr |
C8H8Br2 | |
MFCD00000175 | |
1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776 | |
1,2-bis(bromomethyl)benzene |
Specifications
91-13-4 | |
1.96 | |
MFCD00000175 | |
637159 | |
KGKAYWMGPDWLQZ-UHFFFAOYSA-N | |
1,2-bis(bromomethyl)benzene | |
66665 | |
97% |
92°C to 96°C | |
C8H8Br2 | |
UN3261 | |
1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776 | |
C1=CC=C(C(=C1)CBr)CBr | |
263.96 | |
263.96 | |
o-Xylylene dibromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 202-042-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only