missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Oasis WAX Online Column
The Oasis WAX on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE.
Brand: Waters Corporation 186002508
This item is not returnable.
View return policy
Description
- This column contains the Oasis WAX sorbent, which is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been optimized for fast, simple, and highly selective sample preparation of strong acidic compounds.
- This on-line column is designed to be used with the appropriate Waters narrow-bore analytical column, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry.
- The column features traditional HPLC column fittings and hardware.
Specifications
30 μm | |
No | |
80 Å | |
20 mm |
Solid Phase Extraction Column | |
Spherical | |
1/Ea. | |
2.1 mm |