missing translation for 'onlineSavingsMsg'
Learn More

CellPath OCT Embedding Matrix

Designed for use at -20°C to -30°C

Brand:  CellPath KMA-0100-00A

Product Code. 15212776

This item is not returnable. View return policy

Description

Description

  • Liquid level clearly visible in bottle
  • Highly viscous
  • Freezes rapidly using CryoSpray
  • Easy to use dispenser bottle
  • Medium freezes rapidly with minimal damage by crystal formation and is an ideal support medium for cutting tissue sections
  • Easily removed from microscope slide prior to staining
  • Features a nozzle moulded into the cap
  • Molecular weight of OCT: 94.07g/mol
Specifications

Specifications

125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CellPath OCT Embedding Matrix > Volume: 125mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.