missing translation for 'onlineSavingsMsg'
Learn More

Octadecylamine, 90%, tech., Thermo Scientific Chemicals

£46.73 - £248.24

Chemical Identifiers

CAS 124-30-1
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.52
MDL Number MFCD00008159
InChI Key REYJJPSVUYRZGE-UHFFFAOYSA-N
Synonym octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
PubChem CID 15793
ChEBI CHEBI:63866
IUPAC Name octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10785231
SDS View Documents
Thermo Scientific Acros
129325000
500 g Glass bottle
£46.73
500g
Please to purchase this item. Need a web account? Register with us today!
10665752
SDS View Documents
Thermo Scientific Acros
129320010
1 kg Glass bottle
£73.06
1kg
Please to purchase this item. Need a web account? Register with us today!
10483045
SDS View Documents
Thermo Scientific Acros
129320050
5 kg Plastic drum
£248.24
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Octadecylamine, 90%, tech., Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.