missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltrimethoxysilane, 90%, Tech., Thermo Scientific Chemicals

Product Code. 10300994
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-42-9
Molecular Formula C21H46O3Si
Molecular Weight (g/mol) 374.69
MDL Number MFCD00043060
InChI Key SLYCYWCVSGPDFR-UHFFFAOYSA-N
Synonym octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade
PubChem CID 76486
IUPAC Name trimethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 13°C to 17°C
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 170°C (0.1 mmHg)
Flash Point 139°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. Total octadecyltrimethoxysilanes (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)17Si(OCH3)3
Refractive Index 1.4380 to 1.44
Quantity 100 mL
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble (reacts)
Formula Weight 374.69
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Octadecyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'