missing translation for 'onlineSavingsMsg'
Learn More

Octafluoronaphthalene, 96%, Thermo Scientific Chemicals

£37.80 - £344.00

Chemical Identifiers

CAS 313-72-4
Molecular Formula C10F8
Molecular Weight (g/mol) 272.097
MDL Number MFCD00014307
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
PubChem CID 67564
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490287
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21699.03
1 g
£37.80
1g
Please to purchase this item. Need a web account? Register with us today!
11400297
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21699.06
5 g
£94.90
5g
Please to purchase this item. Need a web account? Register with us today!
11438710
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21699.14
25 g
£344.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water. Solubility in toluene, almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

313-72-4
272.097
JDCMOHAFGDQQJX-UHFFFAOYSA-N
67564
C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F
C10F8
MFCD00014307
octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
1,2,3,4,5,6,7,8-octafluoronaphthalene

Specifications

313-72-4
209°C
MFCD00014307
octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
JDCMOHAFGDQQJX-UHFFFAOYSA-N
1,2,3,4,5,6,7,8-octafluoronaphthalene
67564
96%
84°C to 89°C
C10F8
1915630
Insoluble in water. Solubility in toluene,almost transparent.
C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F
272.097
272.1
Octafluoronaphthalene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 206-239-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only