missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Graduated PET Bottles with IATA-Validated Screw Cap, Pre-Assembled
BIOPHARMA

Catalog Number 16552662
Click to view available options
Capacity (Metric):
1 L
125 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Corning™ Octagonal PET Bottles with IATA-Validated Screw Cap are designed for secure storage of tissue culture media, sera, buffers, and other aqueous solutions

Polyethylene terephthalate (PET) bottles are ideal for storage and sampling of media, biologicals, and other aqueous solutions. The PET resin makes the bottle lightweight, break-resistant and as transparent as glass.

Features:

  • Packaged in two convenient, shrink-wrapped trays within an outer bag to assure cleanliness
  • Break-resistant bottles are an ergonomic and easy to handle alternative to glass
  • Bottles are sterilized to 10−6 SAL and non-pyrogenic
  • Leakproof screw cap is validated against IATA standards for air shipments
  • Molded graduations for accurate measurements
  • USP Class VI compliant bottles are validated non-cytotoxic and non-hemolytic
TRUSTED_SUSTAINABILITY
Capacity (Metric) 250 mL
Capacity (English) 8.5 oz.
Color Clear
Diameter (English) Outer Neck 1.4 in.
Diameter (Metric) Outer Neck 34.7 mm
For Use With (Application) Storage and sampling of media, biologicals and other aqueous solutions
Material PET
No. per Case 48/Cs.
No. per Pack 24/Pk.
Packaging Wrapped Tray
Sterility Sterile
Closure Color Natural
Graduations 50 to 250 mL
Closure Included Yes
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Mouth Narrow
Shape Octagonal
Height (Metric) 133 mm
Product Type Media Storage Bottle
Height (English) 5 in.
Closure Material HDPE
Closure Size 34.7 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'