missing translation for 'onlineSavingsMsg'
Learn More

Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals

Product Code. 11349748
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51777-38-9
Molecular Formula C24H72O10Si11
Molecular Weight (g/mol) 829.765
MDL Number MFCD01310212
InChI Key VLEKPQBXDHFSQO-UHFFFAOYSA-N
Synonym octakis trimethylsiloxy si-lsesquioxane, octakis trimethylsiloxy-t8-silsesquioxane
PubChem CID 71306897
IUPAC Name bis(trimethylsilyl) bis[tris(trimethylsilyloxy)silyl] silicate
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color White
Quantity 100 g
Solubility Information Sparingly Soluble (0.61g/L) (25°C),Calc.
Formula Weight 1130.18
Physical Form Powder
Chemical Name or Material Octakis (trimethylsiloxy)silsesquioxane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'