Learn More
Octanoic acid, 99%, Thermo Scientific Chemicals
Octanoic acid, 99%, C8H16O2, CAS Number-124-07-2, 1-heptanecarboxylic acid, caprylic acid, enantic acid, n-caprylic acid, neo-fat 8, n-octanoic acid, n-octoic acid, n-octylic acid, octoic acid, octylic acid, 1L, 385 to 393mg KOH/g, 98.5% min. (GC), 02, 347, 238.0 deg.C, 100.0, CHEBI:28837
£38.20 - £398.00
Chemical Identifiers
CAS | 124-07-2 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.21 |
MDL Number | MFCD00004429 |
InChI Key | WWZKQHOCKIZLMA-UHFFFAOYSA-N |
Synonym | caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid |
PubChem CID | 379 |
ChEBI | CHEBI:28837 |
IUPAC Name | octanoic acid |
SMILES | CCCCCCCC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10543101
|
Thermo Scientific Acros
129390010 |
1 L | Glass bottle |
£38.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553101
|
Thermo Scientific Acros
129390025 |
2.5 L | Glass bottle |
£69.70
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358573
|
Thermo Scientific Acros
129390251 |
25 L | Metal drum |
£398.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-07-2 | |
144.21 | |
WWZKQHOCKIZLMA-UHFFFAOYSA-N | |
379 | |
octanoic acid |
C8H16O2 | |
MFCD00004429 | |
caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid | |
CHEBI:28837 | |
CCCCCCCC(=O)O |
Specifications
385 to 393mg KOH/g | |
98.5 | |
16.0°C to 16.5°C | |
0.9100g/mL | |
110°C | |
98.5% min. (GC) | |
CH3(CH2)6CO2H | |
MFCD00004429 | |
15, 1767 | |
caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid | |
WWZKQHOCKIZLMA-UHFFFAOYSA-N | |
octanoic acid | |
379 | |
5.8 mPa.s (20°C) | |
99% | |
ACROS Organics |
124-07-2 | |
100.0 | |
Colorless to Yellow | |
238.0°C | |
Authentic | |
C8H16O2 | |
1.4268 to 1.4288 | |
02, 347 | |
0.91 | |
Solubility in water: 0.68g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCCCCC(=O)O | |
144.21 | |
CHEBI:28837 | |
144.21 | |
Liquid | |
Octanoic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-677-5
- Octanoic acid
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only