Learn More
Chemical Identifiers
CAS | 111-64-8 |
---|---|
Molecular Formula | C8H15ClO |
Molecular Weight (g/mol) | 162.657 |
MDL Number | MFCD00000766 |
InChI Key | REEZZSHJLXOIHL-UHFFFAOYSA-N |
Synonym | capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride |
PubChem CID | 8124 |
IUPAC Name | octanoyl chloride |
SMILES | CCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11372196
|
Thermo Scientific Alfa Aesar
L03526.AE |
100 mL |
£21.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382196
|
Thermo Scientific Alfa Aesar
L03526.AP |
500 mL |
£51.05
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOctanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.
Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.
Chemical Identifiers
111-64-8 | |
162.657 | |
REEZZSHJLXOIHL-UHFFFAOYSA-N | |
8124 | |
CCCCCCCC(=O)Cl |
C8H15ClO | |
MFCD00000766 | |
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride | |
octanoyl chloride |
Specifications
111-64-8 | |
0.942 | |
80°C (176°F) | |
C8H15ClO | |
MFCD00000766 | |
635917 | |
capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride | |
REEZZSHJLXOIHL-UHFFFAOYSA-N | |
octanoyl chloride | |
8124 | |
99% |
-63°C | |
195°C | |
Pungent | |
1.435 | |
UN3265 | |
Moisture sensitive | |
Soluble in organic solvents (acetone,chloroform,toluene,THF). Hydrolyzes in water. | |
CCCCCCCC(=O)Cl | |
162.657 | |
162.66 | |
Octanoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
P210-P233-P234-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P333+P313-P362-P374-P390-P501c
H227-H290-H315-H317-H318-H330
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 203-891-6
missing translation for 'rtecsNumber' : RH1570000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only