missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 10489050
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-64-8
MDL Number MFCD00000766
InChI Key REEZZSHJLXOIHL-UHFFFAOYSA-N
Synonym capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
PubChem CID 8124
IUPAC Name octanoyl chloride
SMILES CCCCCCCC(=O)Cl

Specifications

Acidity 1% max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -63.0°C
Density 0.9500g/mL
Boiling Point 195.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H15ClO
Linear Formula CH3(CH2)6COCl
Refractive Index 1.4340 to 1.4360
Quantity 500 g
Beilstein 02, 348
Specific Gravity 0.95
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol, miscible with most common organic solvents, soluble in ether
Molecular Weight (g/mol) 162.66
Viscosity 1.51 mPa.s (20°C)
Formula Weight 162.66
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octanoyl chloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Octanoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'