Learn More
Cytiva Octyl Sepharose™ CL-4B Gel
For use in Hydrophobic Interaction Chromatography (HIC)
Brand: Cytiva 17-0790-01
Code : GE
Additional Details : Weight : 0.33000kg
Description
- Octyl group coupled to cross-linked 4% agarose beads via ether linkage
- Average particle size of 90μm
- Ligand concentration of 40 μmole/ml drained medium
- Stable in all commonly used aqueous buffers and additives such as 8 M urea and 6 M guanidine hydrochloride
- Autoclavable
Specifications
Octyl Sepharose CL-4 B | |
200 mL | |
45 μm-165 μm | |
4% Cross-linked Agarose |
For Hydrophobic Interaction Chromato Graphy | |
Octyl | |
3 to 12 | |
4°C to 30°C, 20% Ethanol or 0.1 M NaOH |
Safety and Handling
- Octyl Sepharose CL-4B, 200 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol