Learn More
Octyltrichlorosilane, 97%, Thermo Scientific Chemicals
£42.40 - £148.00
Chemical Identifiers
CAS | 5283-66-9 |
---|---|
MDL Number | MFCD00000488 |
InChI Key | RCHUVCPBWWSUMC-UHFFFAOYSA-N |
Synonym | trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive |
PubChem CID | 21354 |
IUPAC Name | trichloro(octyl)silane |
SMILES | CCCCCCCC[Si](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10388490
|
Thermo Scientific Acros
216501000 |
100 g |
£42.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10269273
|
Thermo Scientific Acros
216505000 |
500 g |
£148.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5283-66-9 | |
RCHUVCPBWWSUMC-UHFFFAOYSA-N | |
21354 | |
CCCCCCCC[Si](Cl)(Cl)Cl |
MFCD00000488 | |
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive | |
trichloro(octyl)silane |
Specifications
5283-66-9 | |
100.0 | |
1.0700g/mL | |
96°C | |
97% | |
C8H17Cl3Si | |
1.4470 to 1.449 | |
04, III, 1907 | |
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive | |
RCHUVCPBWWSUMC-UHFFFAOYSA-N | |
trichloro(octyl)silane | |
21354 | |
247.67 | |
Liquid |
96.0 | |
Colorless to Yellow | |
233°C (731.0 mmHg) | |
Authentic | |
Glass bottle | |
CH3(CH2)7SiCl3 | |
MFCD00000488 | |
1.07 | |
Solubility in water: reacts | |
CCCCCCCC[Si](Cl)(Cl)Cl | |
247.67 | |
1.9 mPa.s (25°C) | |
97% | |
Octyltrichlorosilane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 226-112-1
- Octyltrichlorosilane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/physician.
RUO – Research Use Only