missing translation for 'onlineSavingsMsg'
Learn More

Implen™ OD600 DiluPhotometer™

A small, easy to use instrument for the determination of cell density and Bradford assays in wavelengths ranging from 590 to 610 nm.

Brand:  Implen™ OD600


Additional Details : Weight : 0.60000kg

Product Code. 16892070

  • £860.49 / Each
Estimated Shipment: 05-11-2024
to see stock.

Description

Description

  • Small, portable spectrophotometer
  • Fast and easy determination of cell density and 600 nm
  • Quantify protein with Bradford protein assay
  • Operating Wavelength: 590 to 610 nm
  • Battery operated, charger included
Specifications

Specifications

<±0.05 A (at 1 A using Neutral Density Filters)
8.5 mm
Semi Micro, Macro Cuvettes (10 mm) or Round Tubes (14 to 16 mm)
Custom LCD
English
40 nm
110/220 V
External Power Adaptor (110/220 V, 50/60 Hz) or Internal Rechargeable NiMH Battery
Cables, Owner's Manual
Optical Density: 0.3 A to 1.99 A
1
No
20 nm
0.3 A to 1.99 A
RS232, USB
LED
180 x 150 x 60 mm
LED
Portable Photometer
590 to 610 nm
Bradford Assays, Bacterial Growth Rates, Colorimetric Protein Assays
6 Button: Power, Blank, Sample, Memory, Reset, Recall/Print
3 sec.
LED
24 Months
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services

For Research Use Only