missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Density Determination Kit for Balances
Use with select OHAUS™ Balances with 0.1 to 1mg resolutions.
Brand: OHAUS™ 80253384
667.67 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Includes: Weighing platform, holder for floating items, holder for non-floating items, beaker stand, beaker, thermometer, forceps
Description
Compatible with:
Models with capacities up to 610g and readability of 0.1 and 1mg
- Discovery
- Explorer
- Adventurer Pro (except AV53x)
- Pioneer
- Explorer
Specifications
Ohaus Balance | |
Weighing platform, holder for floating items, hold | |
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
Density Determination Kits |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction