missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Harvard Trip Balances

Ideal for comparative weighing. OHAUS Harvard Trip Balances with choice of single or double beam models provide many years of maintenance-free, comparative weighing using a known standard.

Brand:  OHAUS™ 80000003

277.44 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 10251012

  • £494.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • With 2000g capacities, 0.1g readabilities, and either a stainless-steel 6 in. (15.2cm) diameter x 0.75 in. deep (1.9cm) pan or 6 in. diameter (15.2cm) plate
  • Angled beams with center-reading poises for excellent visibility of results; sliding, rotating poise for fine adjustment
  • Magnetic damping to slow oscillations and speed weighings
  • Zero-adjust compensator ensures precision and reproducibility
  • Equipped for specific gravity weighing
  • Precision-ground knives, covered self-aligning bearings, cross-braced aluminum-alloy box-end beam, and elliptical check pins
  • Single-beam balances graduated in 0.1g increments; double-beams in 0.1 and 10g increments

Service Plan

Service agreement available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 in.
7 in.
5.9 in.
15 cm
24.1 cm
Stainless Steel
35.5 cm
4.4 lb., 2 kg
15.2 cm
10 x 0.1g (top beam)
22.8 cm
9.5 in.
1450-SD
14 in.
5 Years
Harvard Single Beam Balance
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers