missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ OHS 60 Digital Overhead Stirrer System

System including an OHS 60 Digital Overhead Stirrer, Universal H-Stand, Stirring Shaft with Propeller, and a Double Clamp.

Brand:  Velp Scientifica™ SA20100492

1047.42 GBP valid until 2024-12-31
Use promo code "21913" to get your promotional price.


Product Code. 16575360

This item is not returnable. View return policy

Description

Description

Safety and Resistance in the Lab

  • Resistant sealed housing with IP 54 Protection Rating guarantees durability even in the most aggressive environments
  • Integrated overload, over temperature, and over current safety alarms preserve performance over time by stopping the stirring
  • Safe start and stop knob operation with lock key prevents unintended start up and changes of the working conditions
  • Integrated vibration sensor with different levels of sensitivity will stop the operation and inform users when OHS is stable and properly fixed

Best-in-Class Stirring Power

  • Ideal for demanding applications operating with a range viscosities
  • Electronic controlled, brushless motor stirs with a maximum torque
  • SpeedServo™ torque compensation technology keeps the speed constant automatically adjusting to viscosity changes
  • Two speed gears provide the best performance for both high torque and high volume operations
  • Electronic speed control can manually set to desired rpm
  • Torque remains stable throughout the whole speed range which is a crucial parameter for reproducibility

SmartChuck™ Simplifies Shaft Accomodation

  • Shaft accommodation process can be carried out with one hand and does not require any tools
  • With two simple operations, unlock the protection ring to drive the shaft in and out with maximum ease
  • Opening of the ring during the stirring process will automatically block the analysis and the instrument informs the user for maximum safety

Intuitive and Precise

  • 3.5 in. TFT Display will guide users to precisely set working conditions and get reproducible results
  • Displays main information at a glance together with torque measurement for real-time information on viscosity change with a resolution of 1 Ncm
  • Interact with the OHS Advance from the real-time graph showing speed, torque, and temperature
  • OHS Advance Stirrers work unattended thanks to a timer which can be set from 1 min. to 99.59 hr.
  • Define and program operating methods that are ideal for demanding tasks where ramps and interval operation are required

OHS Advance Connectivity Advantages

  • OHS Advance are the only overhead stirrers to offer the possibility of controlling and monitoring processes through Wi-Fi connection
  • VELP Ermes cloud platform allows to control the OHS Advance from PC, smartphone, and tablet through a simple and intuitive web interface
  • Activate stirring, set a timer, set a method (among those saved on the device), or download and share reports with a click
  • VELP Ermes can manage OHS Advance located in clean-rooms or other controlled environments, without the need of long safety procedures
  • OHS/CotrollerSoft™ software offers all labs a valid alternative with USB connectivity for those who cannot benefit from cloud services
Specifications

Specifications

30 to 2000 rpm
Digital
≤8.5 mm
385 mm
50/60 Hz
Stirring
OHS 60 Digital
IP54
≤50.000 mPas
4.1 kg
40 L
230 V
15.2 in.
230 V, 50/60 Hz
1 to 13 mm
OHS 60 Digital Overhead Stirrer, Universal H-Stand, Stirring Shaft with Propeller, and Double Clamp
Brushless DC
≤60 Ncm
9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers