Learn More
asecos™ Oil Sorbents Cushions
The Oil Sorbents Cusions are water repellant for the safe absorption of oils, petrol, diesel, etc.
£129.32 - £165.36
Specifications
Color White
Material Heavy-duty Cloth
Type Oil Sorbent Pillow
View More Specs
Products 2
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
17856436
missing translation for 'viewDocuments' Promotion Details
asecos™
9933
250.00 mm 250.00 mm
£165.36
Pack of 20
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
17896546
missing translation for 'viewDocuments' Promotion Details
asecos™
9932
460.00 mm 460.00 mm
£129.32
Pack of 10
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibres, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifications
Specifications
White
Heavy-duty Cloth
Oil Sorbent Pillow