Learn More
Kern Sohn OIML Class E2 Compact Test Weight
Single test weight constructed of stainless steel.
Brand: Kern Sohn 316-06
This item is not returnable.
View return policy
Description
- Compact shape
- Polished stainless steel
Specifications
E2 | |
Stainless Steel | |
50 g | |
± 0.1 mg |