Learn More
Kern Sohn OIML Class E2 Compact Test Weight
Single test weight constructed of stainless steel.
Brand:  Kern Sohn 316-09
Encompass_Preferred
 View more versions of this product
Product Code. 17372003
  • £86.82 / Each
Explore more special offers
Description
Description
  • Compact shape
  • Polished stainless steel
Specifications
Specifications
E2
Stainless Steel
500 g
± 0.8 mg