missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleic acid, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
25KG POLY DRUM
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 112-80-1 |
Boiling Point | 360 °C (1,013 hPa) |
Flash Point | 113 °C |
Packaging | Poly Drum |
Molecular Formula | C18H34O2 |
Quantity | 25 kg |
Residual Solvents | Complying |
InChI Key | ZQPPMHVWECSIRJ-MDZDMXLPSA-N |
SMILES | CCCCCCCC\C=C\CCCCCCCC(O)=O |
IUPAC Name | (9E)-octadec-9-enoic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'