Learn More
Novus Biologicals™ Olfactory Marker Protein Overexpression Lysate
Brand: Novus Biologicals™ NBP2-10426
Additional Details : Weight : 0.00970kg
Description
The Olfactory Marker Protein Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_618. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens Olfactory Marker Protein, mRNA. | |
Native |
For Research Use Only