missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oligo(dT)18 Primer

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.

£68.00 - £78.75

Specifications

Product Type Primer
Primer Length 18-mer
Shipping Condition Dry Ice
Concentration 0.5 μg/μL
Primer Oligo dT
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10304690
View Documents
Thermo Scientific™
SO131
60 μL
£68.00
60µL
Please to purchase this item. Need a web account? Register with us today!
10753741
View Documents
Thermo Scientific™
SO132
120 μL
£78.75
120µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR

Related products
Anchored Oligo dT (Cat. No. AB1247)
Random Hexamer Primer (Cat. No. SO142)

Specifications

Specifications

Primer
Dry Ice
Oligo dT
Liquid
18-mer
0.5 μg/μL
cDNA Synthesis
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oligo(dT)18 Primer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.