missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OLIGO™ R3 Reversed - Phase Resin

Product Code. p-7181592 Shop All Applied Biosystems Products
Click to view available options
Quantity:
25 mL
50 mL
1 L
Unit Size:
1 L
25mL
50mL
This item is not returnable. View return policy

Product Code. 18416568

Brand: Thermo Scientific™ 1133903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.

Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.

Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:

  • High performance chromatography resin for analytical to process scale separations
  • High resolution preparative purification at high flow rates
  • Chemical stability—allows aggressive cleaning and sanitization
  • Poly-divinylbenzene particles—yield a robust, easily packable matrix
  • Combines low back pressure, high pH stability, and high capacity

Specifications

Quantity 25 mL
Product Line OLIGO
Product Type R3 Reversed-Phase Resin
Particle Size 30 μm
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Stationary Phase R3
Format Bulk Media
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.