Learn More
Olopatadine hydrochloride, MedChemExpress
MedChemExpress Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.
£68.97 - £136.73
Specifications
Chemical Name or Material | Olopatadine hydrochloride |
---|---|
CAS | 140462-76-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C21H24ClNO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16484357
|
Medchem Express
HY-B0426A |
50 mg |
£136.73
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16418488
|
Medchem Express
HY-B0426A |
10 mg |
£68.97
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.Specifications
Olopatadine hydrochloride | |
White | |
C21H24ClNO3 | |
DMSO : 50 mg/mL (133.74 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (17.84 mM; Need ultrasonic) | |
373.87 | |
99.97% | |
Solid |
140462-76-6 | |
Research | |
ALO4943A KW4679 | |
O=C(O)CC1=CC=C(C/2=C1)OCC3=CC=CC=C3C2=C\CCN(C)C.Cl | |
373.87 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H301∣H400
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.