missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Omega Neotech XP™ Polychloroprene Gloves

Product Code. 15508731
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

The fit and feel of latex, with great chemical protection. Nitritex™ Omega Neotech XP™ Polychloroprene Gloves are ambidextrous, extra length (300mm/12 in.) gloves that are powder-free, latex-free and feature textured fingers and a beaded cuff.

  • Extra length (300mm/12 in.) to provide better forearm protection
  • Polychloropene offers outstanding chemical protection and tactility
  • Beaded cuff provides easier donning and stability on the arm
  • Textured fingers for increased grip
  • Powder-free and latex-free to eliminate Type I latex allergies
  • Easy-access 50-glove dispenser box packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Complies with the requirements of the European Glove Standard EN 420:2003 & EN 374 (Parts 1, 2 & 3:2003)
Classification Class I Medical & Cat 3 PPE
Color Green
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Latex Free Yes
Material Polychloroprene
Powdered Powdered
Size Large
Sterility Non-sterile
Glove Cuff Style Beaded
Packaging Type Dispenser box
Puncture Rating 0.65 AQL
Quantity 50/Bx
Resistant To Chemicals
Series Omega Neotech XP
Length (English) 12 in.
Length (Metric) 300 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'