missing translation for 'onlineSavingsMsg'
Learn More
Omeprazole, 98%, Thermo Scientific™ Product Code.: 15475159

Omeprazole, 98%, Thermo Scientific™

Product Code. 15475159
5g
Click to view available options
Quantity:
1g
5g
This item is not returnable. View return policy

Product Code. 15475159

Brand: Thermo Scientific Alfa Aesar J62860.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A proton pump inhibitor

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.

Solubility
Soluble in water (0.5 mg/ml), DMSO (25 mg/ml), and ethanol (4.5 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73590-58-6
Molecular Formula C17H19N3O3S
Molecular Weight (g/mol) 345.417
MDL Number MFCD00083192
InChI Key SUBDBMMJDZJVOS-UHFFFAOYSA-N
Synonym omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
PubChem CID 4594
ChEBI CHEBI:77260
IUPAC Name 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC

Specifications

Quantity 5g
Merck Index 14,6845
Solubility Information Soluble in water (0.5mg/ml),DMSO (25mg/ml),and ethanol (4.5mg/ml).
Formula Weight 345.42
Percent Purity 98%
Chemical Name or Material Omeprazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP06L037 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberJ62860.06,J62860.03
Lot NumberS26J006 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog NumberJ62860.06,J62860.03,J62860
Lot NumberS02J044 Certificate TypeCertificate of Analysis Date13/06/2023 Catalog NumberJ62860.06,J62860.03
Lot NumberQ21I011 Certificate TypeCertificate of Analysis Date28/04/2022 Catalog NumberJ62860.06,J62860.03,J62860.14
Lot NumberZ03H006 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ62860.06,J62860.03,J62860.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Omeprazole, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.