missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Omeprazole-d3, MedChemExpress
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Omeprazole-d3 (H 16868-d3) is deuterium labeled Omeprazole. Omeprazole, a proton pump inhibitor (PPI), is available for treatment of acid-related gastrointestinal disorders. Omeprazole shows competitive inhibition of CYP2C19 activity with a Ki of 2 to 6 μM. Omeprazole also inhibits growth of Gram-positive and Gram-negative bacteria.
Specifications
Specifications
| Chemical Name or Material | Omeprazole-d3 |
| CAS | 922731-01-9 |
| Purity Grade Notes | Research |
| Physical Form | Powder |
| Molecular Formula | C17H16D3N3O3S |
| Color | Coffee |
| Synonym | H 16868-d3 |
| Solubility Information | DMSO : 125 mg/mL (358.75 mM; Need ultrasonic) |
| SMILES | CC(C(OC)=C(C)C=N1)=C1CS(C2=NC3=CC=C(OC([2H])([2H])[2H])C=C3N2)=O |
| Molecular Weight (g/mol) | 348.43 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction