Learn More
Specac™ Omni-Cell™ Fused Silica Mull Windows Circular 25mm Diameter
Brand: Specac™ GS01838
This item is not returnable.
View return policy
Description
Fused Silica circular mull cell windows for use in the Omni-Cell GS01800. 25mm diameter and 2mm thick.Specifications
Omni-Cell GS01800 | |
2 mm thick | |
25 mm |