Learn More
Diba Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces
Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces feature PEEK™ column threads which offer outstanding durability
Brand: Diba 006EZ-06-25-FF
This item is not returnable.
View return policy
Description
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
4 to 40°C | |
Liquid Chromatography Column | |
10 and 30 μm | |
250 mm | |
40°C |
6.6 mm | |
900 psi | |
7.5 mL | |
Borosilicate Glass, PTFE, Polyethylene, FKM/FPM | |
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF Polypropylene fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces
Spot an opportunity for improvement?Share a Content Correction