Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
Brand:  Diba 006EZS-35-15-FF
Encompass_Preferred
 View more versions of this product
Product Code. 15825494
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • PEEK™ column threads offer outstanding durability
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Specifications
4 to 40°C
Liquid Chromatography Column
10 and 30 μm
150 mm
40°C
35 mm
150 psi
115.5 mL
Borosilicate Glass, PTFE, Chemraz™
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces