Learn More
Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns feature a range of porosities for your media and chemistry
Brand: Diba 006FR-25-25-PE
This item is not returnable.
View return policy
Specifications
Chromatography Column Frit, 25mm dia, 25 μm, Polyethylene; 20/pk | |
25 mm | |
25 μm |
Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns | |
Liquid Chromatography Column | |
Polyethylene |