Learn More
Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns feature a range of porosities for your media and chemistry
Brand: Diba 006FR-25-50
This item is not returnable.
View return policy
Specifications
Chromatography Column Frit, 25mm dia, 50 μm, PTFE; 20/pk | |
25 mm | |
50 μm |
Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns | |
Liquid Chromatography Column | |
PTFE |