missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Omnifix Solo, Eccentric Luer-tip, 3-part Syringe

Use the B Braun™ Omnifix Solo, Eccentric Luer-tip, 3-part Syringe for patient care when for slow aspiration and injection are necessary.

Specifications

Certifications/Compliance EN ISO 7886-1, ISO 10993 resp. DIN EN 30993
Material Polypropylene (Plunger), Synthetic Rubber (Stopper)
Color Natural
Description High contrast black graduation
For Use With (Application) For slow aspiration and injection
View More Specs

Description

Description

  • Disposable
  • High contrast black graduations
  • Eccentric Leur-tip
Specifications

Specifications

EN ISO 7886-1, ISO 10993 resp. DIN EN 30993
Natural
For slow aspiration and injection
Eccentric Luer-tip 3-part Syringe
100/Pk.
Yes
Polypropylene (Plunger), Synthetic Rubber (Stopper)
High contrast black graduation
Yes
Single-use
Eccentric Luer Tip
Sterile
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 5 Years

SDS
Documents

Documents

Product Certifications