Learn More
Fisherbrand™ omniPAGE Blot Insert for omniPAGE Blot MaxiWAVE Vertical Electrophoresis System
Ensures to maintain even contact between the gel and membrane.
Brand:  Fisherbrand™ VS20BI
Product Code. 12328037
  • £387.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Used for Western blotting of proteins
  • Cassettes open architecture ensures the maximum blot area allowing direct transfer of current
Specifications
Specifications
OmniPAGE Blot MaxiWAVE System
Blot Insert
Three cassettes and 12 fibre pads
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ omniPAGE Blot Insert for omniPAGE Blot MaxiWAVE Vertical Electrophoresis System > omniPAGE Blot Insert