Learn More
Onartuzumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen MA541934
Product Code. 17768553
  • £415.00 / 100µg
Explore more special offers
Description
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Onartuzumab has been used in trials studying the treatment of Neoplasms, Lung Cancer, Glioblastoma, Gastric Cancer, and Colorectal Cancer, among others.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Onartuzumab
Recombinant Monoclonal
Unconjugated
MetMAb; OA-5D5; PRO 143966
Human MET/RCCP2
100 μg
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
2 mg/mL
PBS with no preservative; pH 7.4
Human
Protein L
RUO
Chemical
Antibody
IgG1 F(ab) κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Onartuzumab Recombinant Human Monoclonal Antibody, Invitrogen™ > 100 μg; Unconjugated