missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Oncovirus Epitope Microarray

Oncovirus Epitope Microarray with 4463 epitope specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.036.001

Product Code. 16307778

  • £515.00 / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Microarray Content: 3653 linear B-cell epitopes of human oncoviruses from the Immune Epitope Database including 7 citrullinated peptides and the corresponding arginine variants
  • Database Epitope Lengths: 3 to 50 aa
  • Peptide Lengths: 3 aa to 20 aa depending on the epitope length; longer epitopes were translated into overlapping 20 aa peptides
  • Number of Peptides: 4463 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEGMA/PMMA
1 Slide
1 mm
Transparent
Individual barcode for each microarray
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Microscopic Glass
Antibody validation, epitope mapping, serum screening, biomarker discovery
75.4 x 25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers