missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ONE Broth-Salmonella Base

Product Code. p-6980032
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
This item is not returnable. View return policy

Product Code. 15832347

Brand: Thermo Scientific™ CM1091T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enrich and recover Salmonella spp. in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base.

Enrich and recover Salmonella spp. from food samples in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base, when prepared with Thermo Scientific Oxoid ONE Broth-Salmonella Selective Supplement (SR0242).

ONE Broth-Salmonella Base is a highly nutritious and selective medium that allows recovery of Salmonella species within 16 to 20 hours. ONE Broth-Salmonella Base requires only a single enrichment medium step. When followed by inoculation onto Thermo Scientific™ Brilliance™ Salmonella Agar and the Thermo Scientific Oxoid Salmonella Latex Test, it forms the Oxoid™ Salmonella Precis™ method which reduces time to result over conventional culture methods. The Oxoid Salmonella Precis method is AFNOR validated.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description ONE Broth-Salmonella Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 5 kg
Species Salmonella
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.