Learn More
Hamilton™ One Hub Assembled
One Hub Assembled with 24 in. of Tubing 22 gauge
Brand:  Hamilton™ 8892-01
Product Code. 11543114
  • £117.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Specification Sheet
Fitting MaterialCTFE
Tubing MaterialPTFE
Gauge22 gauge
Length24 inches
Sales Quantity1 each
Fitting Type1/4 in.-28
Number of Hubsone
Specifications
Specifications
1/4 in.-28 in.
24 in.