missing translation for 'onlineSavingsMsg'
Learn More
3M™ One Touch™ Pro Earplug Dispenser Product Code.: 15513483
Estimated Shipment: 11-04-2025
to see stock.
Add to basket

3M™ One Touch™ Pro Earplug Dispenser

Product Code. 15513483
Color: Blue
Click to view available options
Color: Blue
This item is not returnable. View return policy

Product Code. 15513483

Brand: 3M™ 7100064963

Estimated Shipment: 11-04-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

The 3M™ One Touch™ Pro Earplugs dispenser offers convenient access to workers in a busy workplace. A simple twist accurately and consistently dispenses one earplug at a time. This reusable free-standing or wall hanging dispenser can be re-stocked with One Touch™ refills of the most popular 3M™ E-A-R™ Earplugs.

  • Accurately dispenses one earplug at a time
  • Dispenses earplug directly into hand
  • Durable ABS plastic for tough environments
  • Free-standing or wall-mounted, could be positioned inside or outside
  • Shields earplugs from sun, wind and rain when dispensing earplugs outside
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Certifications/Compliance CE Approved
For Use With (Equipment) For use with E-A-R™ disposable ear plug refills
Product Type Ear Plug Dispenser
Material ABS plastic
Mounting Wall Mountable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ One Touch™ Pro Earplug Dispenser >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.