missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Red, 594

Product Code. 18377022 Shop All Bio Techne Products
Click to view available options
Quantity:
20 Assays
Unit Size:
20 assays
This item is not returnable. View return policy

Product Code. 18377022

Brand: Novus Biologicals™ NBP31195920ASSAYS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for in situ apoptosis detection of tissue samples (Paraffin embedding, freezing and ultrathin section) and cells (Cell smears, cell climbing smears).

TRUSTED_SUSTAINABILITY

Specifications

Description OneStep TUNEL
Content And Storage Store at -20 C.
Quantity 20 Assays
For Use With (Application) Flow Cytometry, Fluorescence Imaging
Product Type Apoptosis Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.