missing translation for 'onlineSavingsMsg'
Learn More

ONO 2506, Tocris Bioscience™

Product Code. 30001659
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits S-100β synthesis; neuroprotective

Inhibits S-100β synthesis in activated cultured astrocytes. Prevents delayed infarct expansion 24 hours after permanent middle cerebral artery occlusion (pMCAO) in rats; also exhibits neuroprotective effects in mouse models of Parkinson's disease and Alzheimer's disease.

Chemical Identifiers

CAS 185517-21-9
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
InChI Key YCYMCMYLORLIJX-SNVBAGLBSA-N
Synonym arundic acid, 2r-2-propyloctanoic acid, r-2-propyloctanoic acid, cereact, proglia, acide arundique, acido arzndico, acidum arundicum, r---arundic acid, arundic acid inn
PubChem CID 208925
IUPAC Name (2R)-2-propyloctanoic acid
SMILES CCCCCCC(CCC)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 186.29
Percent Purity >98%
Chemical Name or Material ONO 2506
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'