missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant, Anti-glare Medical Mask

Be prepared for long hours of wear with this anti-glare, anti-fog medical mask complete with a comfortable foam band and anti-reflective tape.

Brand:  Kolmi Hopen™ M34101MN-30

Additional Details : Weight : 1.52000kg

Product Code. 17740199

  • £7.30 / Pack of 40
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Protects environment from droplets from wearer
  • Provides protection against biological fluids under pressure of 16 kPa
  • Non-irritating inner layer designed to reduce moisture and wick away sweat
  • Used by healthcare workers to reduce risk of infection or in the operating room
  • Anti-reflective black tape to prevent glare in operating room
  • Foam band to reduce fogging of glasses
Specifications

Specifications

Blue
Polypropylene
(EU) 2017/745, ISO 9001, ISO 13485, EN 14683
Medical
Ties
175 x 95 mm
Yes
40 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers